Specifications
- Description:Microtome blades, low profile
- Cat. No.:103790-914
Specifications
About this item
Tek-Select® Microtome Blades provide a quality cut with a long-lasting, ultra-sharp edge.
- High profile and low profile formats
- Durable construction for a quality cut
- Long-lasting edge
- Ultra-sharp edges feature a special resin coating
Tek-Select® Microtome Blades feature an ultra-sharp edge with a special resin coating. Available in high profile or low profile formats, the blades are consistently sharp from pack to pack. Packaged 50 blades per box.