ContactUsLinkComponent
Tek-Select® Fan-Pad Mini-Formalin Absorbent Pad, IMEB
Tek-Select® Fan-Pad Mini-Formalin Absorbent Pad, IMEB
Catalog # 103783-056
Supplier:  IMEB INC MS
undefined
Tek-Select® Fan-Pad Mini-Formalin Absorbent Pad, IMEB
Catalog # 103783-056
Supplier:  IMEB INC MS
Supplier Number:  FAN-55
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Formalin absorbent pads
  • L×W:
    11×10.75"
  • Cat. No.:
    103783-056
Specifications
About this item
Tek-Select® Fan-Pad mini formalin absorbent are specially treated to absorb formalin and formaldehyde.
  • Pads absorb formalin and formaldehyde
  • Specially treated surfaces
  • 55 sheets per roll
Tek-Select® Fan-Pad mini formalin absorbent pads ensure that formalin products are absorbed and harmful vapors neutralized. Pads can be disposed in regular trash receptacles.