Order Entry
Puerto Rico
ContactUsLinkComponent
Bacillus anthracis Poly-gamma-D-Glutamic Acid
Bacillus anthracis Poly-gamma-D-Glutamic Acid
Catalog # 103007-710
Supplier:  Anaspec Inc
CAS Number:  
Bacillus anthracis Poly-gamma-D-Glutamic Acid
Catalog # 103007-710
Supplier:  Anaspec Inc
Supplier Number:  AS-63756
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Bacillus anthracis
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    DPGA Peptide|Bacillus anthracis capsule sequence
  • Protein/Peptide Name:
    Poly-gamma-D-Glutamic Acid
  • Purity:
    95%
  • Molecular Weight:
    2099 Da
  • Sequence:
    Ac-(g-e)15-C-NH2
  • Cat. No.:
    103007-710
  • Supplier no.:
    AS-63756

Specifications

About this item

This peptide is a poly-gamma-D-glutamic acid (DPGA)10 construct that relates to the sequence of the Bacillus anthracis capsule which is composed of gamma DPGA. gamma DPGA is an essential virulence factor of B. anthracis. The capsule inhibits innate host defense through its antiphagocytic action. gamma DPGA is a poor immunogen, but when bound to a carrier protein, it elicits serum antibodies.

References: Kubler-Kielb, J. et al. Infect. Immun. 74, 4744 (2006).