Order Entry
Puerto Rico
ContactUsLinkComponent
Hepatitis Virus C NS3 Protease Inhibitor 2
Hepatitis Virus C NS3 Protease Inhibitor 2
Catalog # 102996-708
Supplier:  Anaspec
CAS Number:  
Hepatitis Virus C NS3 Protease Inhibitor 2
Catalog # 102996-708
Supplier:  Anaspec
Supplier Number:  AS-25346
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Hepatitis virus
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    HCV inhibitor Peptide
  • Protein/Peptide Name:
    Hepatitis Virus C NS3 Protease Inhibitor 2
  • Purity:
    95%
  • Molecular Weight:
    913.1 Da
  • Sequence:
    Ac-DE-Dif-E-Cha-C
  • Cat. No.:
    102996-708
  • Supplier no.:
    AS-25346

Specifications

About this item

A potent peptide inhibitor of Hepatitis Virus C NS3 protease.

Ref: Ingallinella, P. et al. Biochem. 37, 8906 (1998); Ferreon, J. et al. J. Biol. Chem. 280, 20483 (2005).

Product Citations:
Gutierrez-Grobe, Y. et al. (2011). Biliverdin and Heme Oxygenase Antibiral Activity Against Hepatitis C Virus. Annals Hepatology 10, 105.

Nam, SY. et al. (2011). Abdominal visceral adipose tissue volume is associated with increased risk of erosive esophagitis in men and women. Eur J Gastroenterology Hepatology 23, 197.

Zhu, Zhaowen, et al. (2010). Biliverdin inhibits hepatitis C virus NS3/4A protease activity: Mechanism for the antiviral effects of heme oxygenase? Hepatology DOI: 10.1002/hep.23921