Order Entry
Puerto Rico
ContactUsLinkComponent
Isotonic NaCl Prefilled Syringes, SAI Infusion Technologies
Isotonic NaCl Prefilled Syringes, SAI Infusion Technologies
Catalog # 76408-390
undefined
Isotonic NaCl Prefilled Syringes, SAI Infusion Technologies
Catalog # 76408-390
Supplier Number:  SC10-10

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Sterility:
    Sterile
  • Volume:
    10 ml
  • Description:
    Isotonic NaCl prefilled syringes
  • Cat. No.:
    76408-390

Specifications

About this item

Prefilled syringes for flushing, locking or sampling through an indwelling catheter or port system. This prefilled option can save you time, space and money.

  • Saline (0.9%) filled syringes with caps
  • 10 ml syringe filled to 10 ml
  • Sterile syringes with caps
  • No need to unwrap individual syringes
  • No need to store physiologic containers

Size, volume and needle customizations available.

Caution: For animal use only.