ContactUsLinkComponent
Sani Professional® No-Rinse Sanitizing Multi-Surface Wipes
Sani Professional® No-Rinse Sanitizing Multi-Surface Wipes
Catalog # 76410-930
undefined
Sani Professional® No-Rinse Sanitizing Multi-Surface Wipes
Catalog # 76410-930
Supplier Number:  NICP56784
Specifications
  • Color:
    White
  • Cat. No.:
    76410-930
  • Packaging:
    95 Wipes/container
  • Scent:
    Unscented
Specifications
About this item
Single-use, disposable wipes for easy cleaning and sanitizing. Wipes are pre-measured with sanitizing solution to prevent cross-contamination and protect against foodborne pathogens like E. coli and salmonella.
  • Eliminate the need for buckets and dirty rags
  • Chemical compound: Alkyl dimethyl benzyl ammonium chloride; didecyl dimethyl ammonium chloride
  • Convenient way to sanitize, one wipe at a time
It provides a convenient way to sanitize, one wipe at a time. Use on chairs, countertops, tables, and foodservice surfaces. Ideal for use on hard surfaces, such as stainless steel.
Certifications: EAR99.