Valve for ASXPRESS PLUS, 6-Port, PerkinElmer
Catalog # 10139-508
Supplier: PerkinElmer
Specifications
- Description:6-Port valve for ASXPRESS PLUS
- Cat. No.:10139-508
- Supplier no.:N0777575
Specifications
About this item
This 6-Port valve, designed for use with aqueous applications, is specifically designed for the ASXPRESS® PLUS.
- 1.52 mm diameter ports
- Labels included
The valve, manufactured from polyphenylene sulfide (PPS), contains six ports, each 1.52 mm in diameter. Labels for the ports are included.