ContactUsLinkComponent
Viton Drain Pump Tubing and Connector Kit, PerkinElmer
Viton Drain Pump Tubing and Connector Kit, PerkinElmer
Catalog # 10136-458
Supplier:  PerkinElmer
Viton Drain Pump Tubing and Connector Kit, PerkinElmer
Catalog # 10136-458
Supplier:  PerkinElmer
Supplier Number:  N0777455
Specifications
  • Description:
    Viton drain pump tubing and connector kit
  • Cat. No.:
    10136-458
  • Supplier no.:
    N0777455
Specifications
About this item
This Drain Pump Tubing and Connector Kit, for oils applications, contains Viton® tubing and connectors for the peristaltic pumps in the ASX-260, ASX-520, ASX-520HS, EXR-8, XLR-8, ASX-1400, ASX-1600, and APS-1650.
Enough tubing and connectors are included for two replacements.