ContactUsLinkComponent
Zero Dilution Glass Outer Liner, 2 mm, PerkinElmer
Zero Dilution Glass Outer Liner, 2 mm, PerkinElmer
Catalog # 97019-182
Supplier:  PerkinElmer
undefined
Zero Dilution Glass Outer Liner, 2 mm, PerkinElmer
Catalog # 97019-182
Supplier:  PerkinElmer
Supplier Number:  N1011445
Specifications
  • Description:
    Zero dilution glass outer liner
  • Cat. No.:
    97019-182
  • Inner diameter:
    2 mm
  • Supplier no.:
    N1011445
Specifications
About this item
The Zero Dilution Glass Outer Liner is ideal for trace HS work.
  • Compatible with the 480, 580, 680 and older Clarus GC models
  • Not compatible with Clarus 590/690 capillary injector
Inlet liners for split injection have mixing chambers with tortuous flow paths to allow full vaporization of the sample. Deactivating the surface of these liners prevents active compounds from degrading. Packing the liner with wool will trap non-volatile residue and prevent column contamination when analyzing dirty samples. Inlet liners for splitless injection are generally designed as straight tubes, although new designs such as the gooseneck will help contain the sample in the injector. Packing these liners with wool will also help trap non-volatile residue and prevent column contamination.