PolyScience® Replacement Air Filter, PerkinElmer
Catalog # 10139-242
Supplier: PerkinElmer
Specifications
- Description:PolyScience® replacement air filter
- Environmentally Preferable:
- Cat. No.:10139-242
- Supplier no.:N0777095
Specifications
About this item
This replacement air filter is designed for use with the PolyScience® WhisperCool™ refrigerated chiller in ICP-MS and ICP-OES instruments. This 13×14" air filter with frame is fully washable and removes quickly for easy cleaning.
Filter is designed specifically for use with the PolyScience® WhisperCool™ refrigerated chillers.