ContactUsLinkComponent
SeaSpray™ Glass Nebulizer for NexION, Elan®, and Optima™ Instruments, PerkinElmer
SeaSpray™ Glass Nebulizer for NexION, Elan®, and Optima™ Instruments, PerkinElmer
Catalog # 10139-472
Supplier:  PerkinElmer
undefined
SeaSpray™ Glass Nebulizer for NexION, Elan®, and Optima™ Instruments, PerkinElmer
Catalog # 10139-472
Supplier:  PerkinElmer
Supplier Number:  N0777484
Specifications
  • Description:
    SeaSpray™ glass nebulizer for NexION, Elan®, and Optima™ instruments
  • Cat. No.:
    10139-472
  • Supplier no.:
    N0777484
Specifications
About this item
The SeaSpray™ nebulizer is designed for use with ELAN® 5000, 6x00, 9000, and DRCs and NexION® 300 and 350 ICP-MS series instruments, as well as Optima® 3x00 RL ICP-OES series instruments.
  • Glass nebulizer
  • 0.4 ml/min. uptake rate
The SeaSpray™ concentric nebulizer is ideal for samples with high dissolved solids and offers outstanding nebulization efficiency for trace level analysis. It offers freedom from clogging while nebulizing solutions near the limit of solubility of most mineral salts and conferring significant sensitivity gains. They are supplied with the EzyLok Argon connector.