Clorox® Healthcare® Fuzion™ Cleaner Disinfectant
76408-392
- Description:Fuzion™ Cleaner Disinfectant spray
- Size:32 oz
- Environmentally Preferable:
- Ingredients:Sodium hypochlorite
- Type:Spray disinfectant
- Chemical/physical specifications:Sodium hypochlorite-based
- Cat. No.:76408-392
- Scent:Unscented
Bleach-based cleaner disinfectant.
- A multi-surface cleaner with bleach that’s fast acting and ready to use
- Ideal for use in healthcare and acute care facilities, physician and dental offices, athletic centers, schools and long-term care
- Kills C. difficile spores in 2 minutes, the fastest kill time available based on comparison of EPA master labels as of March 2018
- One minute kill time for 52 types of bacteria, viruses and fungi, including Norovirus, MRSA, TB, VRE, and Parvovirus
- This powerful bleach cleaner contains a neutralizer, which breaks down the bleach to significantly reduce residue and reduce lingering bleach odor
- Broad surface compatibility on surface materials, covers twice as much area with each spray, when compared with traditional swirl nozzles
- Does not cause long term surface damage such as staining, cracking or corroding
- No PPE required
Use in Healthcare settings including hospitals, medical facilities, dental offices, veterinary offices and long term care facilities.
Applicable Material: Ceramic; Enamel; Fiberglass; Finished/Sealed Wood; Laminates; Metal; Plastic; Sealed/Engineered Stone; Vinyl
Dirt Types: Bacteria; Dust; Germs; Grease; Grime; Oil-Based Soil; Organic Matter; Smoke Film; Soap Scum; Water-Based Soil