ContactUsLinkComponent
Avantor® Hichrom, Frit Assembly
Avantor® Hichrom, Frit Assembly
Catalog # 76399-148
Supplier:  Avantor
Avantor® Hichrom, Frit Assembly
Catalog # 76399-148
Supplier:  Avantor
Supplier Number:  HI-119
Specifications
  • Description:
    Frit assembly, 0.25×1.5 mm
  • Material:
    PEEK
  • Cat. No.:
    76399-148
Specifications
About this item
The frit assembly is made of PEEK.