UV Lamps for Millipore Lab Water Systems, Avidity Science™
Supplier: AVIDITY SCIENCE
Certificates
About this item
These UV lamps are compatible with several lab water systems.
Available for following Millipore Lab Water Units: MilliQ Academic, MilliQ Advantage A10, MilliQ Biocel, MilliQ Element, MilliQ Gradient, MilliQ Integral, MilliQ Reference, MilliQ Synthesis, Elix, Super-Q Plus.