Order Entry
Puerto Rico
ContactUsLinkComponent
Special Optical Glass Macro Cylindrical Cell with PTFE Stopper, PerkinElmer
Special Optical Glass Macro Cylindrical Cell with PTFE Stopper, PerkinElmer
Catalog # 45603-834
Supplier:  PerkinElmer
Special Optical Glass Macro Cylindrical Cell with PTFE Stopper, PerkinElmer
Catalog # 45603-834
Supplier:  PerkinElmer
Supplier Number:  B0631094

Specifications

  • Description:
    Special optical glass macro cyl. Cell
  • Lightpath:
    100 mm
  • Cat. No.:
    45603-834
  • Pathlength:
    100 mm
  • Supplier no.:
    B0631094

Specifications

About this item

The special optical glass macro cylindrical cell has a volume of 28 ml, a light path of 100 mm, and an outside depth of 102.5 mm.

Cylindrical cells are used with spectrophotometers that have a larger circular light beam. Each cell has an outside diameter of 22 mm and an inside diameter of 19 mm. Each package contains one cell and two windows.