ContactUsLinkComponent
Rubidium (Rb) System 2 Electrodeless Discharge Lamp, PerkinElmer
Rubidium (Rb) System 2 Electrodeless Discharge Lamp, PerkinElmer
Catalog # 45603-430
Supplier:  PerkinElmer
undefined
Rubidium (Rb) System 2 Electrodeless Discharge Lamp, PerkinElmer
Catalog # 45603-430
Supplier:  PerkinElmer
Supplier Number:  N3050664
Specifications
  • Description:
    Rubidium (Rb) system 2 edl
  • Cat. No.:
    45603-430
  • Supplier no.:
    N3050664
Specifications
About this item
System 2 electrodeless discharge lamp for the detection of elemental Rubidium (Rb). System 2 Electrodeless Discharge Lamps (EDLs) provide the optimized spectral output needed to get the maximum performance from , PerkinElmer® Atomic Absorption spectrometers.
Note: PinAAcle™ instruments require the purchase of an EDL driver assembly, part number 03030997, for each of the lamp positions to be used. Electrodeless Discharge Lamps for greater brightness (EDLs) are typically much brighter and, in some cases, provide better sensitivity than comparable HCLs. EDLs are preferred for certain volatile elements. They offer better precision and lower detection limits for analyses that are “noisy” due to weak hollow cathode emission. Electrodeless Discharge Lamps (EDLs) are designed to be physically interchangeable with Intensitron Hollow Cathode Lamps. Their power supplies are built into all the PinAAcle™ series of atomic absorption spectrometers. Use of EDLs with those instruments requires only the purchase of an EDL driver assembly for each of the lamp positions to be used.