ContactUsLinkComponent
Non-Coded Lumina Lamp Plug Adapter, PerkinElmer
Non-Coded Lumina Lamp Plug Adapter, PerkinElmer
Catalog # 97013-406
Supplier:  PerkinElmer
undefined
Non-Coded Lumina Lamp Plug Adapter, PerkinElmer
Catalog # 97013-406
Supplier:  PerkinElmer
Supplier Number:  N3050198
Specifications
  • Description:
    Non-coded lumina lamp plug adapter
  • Cat. No.:
    97013-406
  • Supplier no.:
    N3050198
Specifications
About this item
Allows Lumina™ HCLs (N305XXXX series, 4-pin) to be used with , PerkinElmer atomic absorption instruments with non-coded lamp connectors (9-pin).
PerkinElmer also offers lamp adapters that are coded to specific element(s).