SW® PowerForm® PF-95GW Dual-Color Green/White 5.9 mil Sustainable Skin-Improving Nitrile Exam Gloves
Supplier: SW Safety Solutions
Certificates
About this item
Experience EnerGel® infused gloves, designed to hydrate skin and protect against long-term glove usage, with sustainable EcoTek® technology.
- EnerGel® hand conditioning technology hydrates and soothes skin; proven to prevent many skin ailments
- EcoTek® sustainable technology
- Breach Alert® visual detection technology provides durable 2-ply construction and contrasting inner color
- Proven resistance to fentanyl and gastric acid for over 4 hours to replicate hazardous settings and overdose conditions
- Fully textured for secure grip
- Silicone-free for critical environments
- 1.5 AQL exceeds laboratory standards
EcoTek® gloves tested to ASTM D5526-12 standard test method for determining anaerobic biodegradation of plastic materials under accelerated landfill conditions. Results are 92.6% biodegradation in 60% solids landfill at 945 days. Future results cannot be predicted/extrapolated. For additional information on the test reports, contact [email protected].
Certifications: WBENC Women Owned; NCMSDC Minority Business; Small Business Enterprise; SHA Dermatologically Approved; EAC TP TC 019/2011; CE 0493; CE EN455.
Ordering information: Glove sizes are as follows; XS (5.5 to 6), S (6.5 to 7), M (7.5 to 8), L (8.5 to 9), XL (9.5 to 10), XXL (10.5 to 11).
Specifications
- Catalog No:
- 76406-612
- 76406-614
- 76406-616
- 76406-618
- 76406-620
- 76406-622
- Model:
- PF-95GW
- PF-95GW
- PF-95GW
- PF-95GW
- PF-95GW
- PF-95GW
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Environmentally Preferable:
- Safer Human and Environmental Health, Sustainable Packaging
- Safer Human and Environmental Health, Sustainable Packaging
- Safer Human and Environmental Health, Sustainable Packaging
- Safer Human and Environmental Health, Sustainable Packaging
- Safer Human and Environmental Health, Sustainable Packaging
- Safer Human and Environmental Health, Sustainable Packaging
- Disposable/Reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Powdered/Powder-free:
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Finish:
- Fully textured
- Fully textured
- Fully textured
- Fully textured
- Fully textured
- Fully textured
- Cuff Style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Cuff Length:
- 242 mm (9.5")
- 242 mm (9.5")
- 242 mm (9.5")
- 242 mm (9.5")
- 242 mm (9.5")
- 242 mm (9.5")
- Cuff Thickness:
- 0.10 mm (3.8 mil)
- 0.10 mm (3.8 mil)
- 0.10 mm (3.8 mil)
- 0.10 mm (3.8 mil)
- 0.10 mm (3.8 mil)
- 0.10 mm (3.8 mil)
- Finger Thickness:
- 0.15 mm (5.9 mil)
- 0.15 mm (5.9 mil)
- 0.15 mm (5.9 mil)
- 0.15 mm (5.9 mil)
- 0.15 mm (5.9 mil)
- 0.15 mm (5.9 mil)
- Palm Thickness:
- 0.13 mm (4.9 mil)
- 0.13 mm (4.9 mil)
- 0.13 mm (4.9 mil)
- 0.13 mm (4.9 mil)
- 0.13 mm (4.9 mil)
- 0.13 mm (4.9 mil)
- FDA Regulated:
- +
- +
- +
- +
- +
- +
- Tensile Strength:
- 18 Mpa
- 18 Mpa
- 18 Mpa
- 18 Mpa
- 18 Mpa
- 18 Mpa
- Ultimate Elongation:
- 500%
- 500%
- 500%
- 500%
- 500%
- 500%
- EN Norm:
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- Food Contact Approved:
- +
- +
- +
- +
- +
- +
- ASTM No.:
- D6319-19; D6978-05 (2019); F1671-13; F739-12
- D6319-19; D6978-05 (2019); F1671-13; F739-12
- D6319-19; D6978-05 (2019); F1671-13; F739-12
- D6319-19; D6978-05 (2019); F1671-13; F739-12
- D6319-19; D6978-05 (2019); F1671-13; F739-12
- D6319-19; D6978-05 (2019); F1671-13; F739-12
- Environmental Certification:
- FSC Certified (BV-COC-170864), C2C Material Health Certified
- FSC Certified (BV-COC-170864), C2C Material Health Certified
- FSC Certified (BV-COC-170864), C2C Material Health Certified
- FSC Certified (BV-COC-170864), C2C Material Health Certified
- FSC Certified (BV-COC-170864), C2C Material Health Certified
- FSC Certified (BV-COC-170864), C2C Material Health Certified