Order Entry
Puerto Rico
ContactUsLinkComponent
Kalrez® Small Seal O-Ring, High Temperature, Size 009, PerkinElmer
Kalrez® Small Seal O-Ring, High Temperature, Size 009, PerkinElmer
Catalog # 97012-746
Supplier:  PerkinElmer
undefined
Kalrez® Small Seal O-Ring, High Temperature, Size 009, PerkinElmer
Catalog # 97012-746
Supplier:  PerkinElmer
Supplier Number:  M0415360

Specifications

  • Description:
    Kalrez Small seal O-ring, high temperature, Size 009
  • Material:
    Kalrez
  • Cat. No.:
    97012-746
  • Supplier no.:
    M0415360

Specifications

About this item

Designed for use in the TurboMatrix Upper Seal Assembly and at high temperatures (above 225 °C).

  • I.Ø×O.Ø: 0.208×0.348"

Compatible with the following PerkinElmer instruments: TurboMatrix 650 ATD, TurboMatrix 150 ATD, Autosystem, TurboMatrix 100 TD, TurboMatrix 300 TD, Autosystem XL, TurboMatrix 350 ATD.