- Description:Beryllium (Be) Lumina™ HCL
- Cat. No.:45603-266
- Supplier no.:N3050110
Single Element Lumina™ Hollow Cathode Lamp (HCL) for the detection of elemental Beryllium (Be). Series N30501XX Lumina™ 2" (50 mm) diameter single-element lamps are designed to be used with our PinAAcle™ and AAnalyst™ Atomic Absorption spectrometer series of instruments.
Please note that this element requires the use of a nitrous oxide-acetylene flame. Every genuine lamp is designed for use with and tested on our spectrometers to assure compatibility and the highest performance. We offer a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ Hollow Cathode Lamp (HCL), we produce lamps with larger internal volume so that a greater supply of fill gas at optimum pressure is available.
The larger the lamp, the greater the inert gas volume - and the longer the lamp lifetime. Lumina™ Hollow Cathode Lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable.