ContactUsLinkComponent
PC³-LT Peltier Cooler Organics Sample Introduction Kit for Optima™ 2X00/3X00/4X00/5X00/7X00 DV/8X00, PerkinElmer
PC³-LT Peltier Cooler Organics Sample Introduction Kit for Optima™ 2X00/3X00/4X00/5X00/7X00 DV/8X00, PerkinElmer
Catalog # 10138-280
Supplier:  PerkinElmer
undefined
PC³-LT Peltier Cooler Organics Sample Introduction Kit for Optima™ 2X00/3X00/4X00/5X00/7X00 DV/8X00, PerkinElmer
Catalog # 10138-280
Supplier:  PerkinElmer
Supplier Number:  N0777356
Specifications
  • Description:
    PC³-LT Organics sample intro kit
  • Environmentally Preferable:
  • Cat. No.:
    10138-280
  • Supplier no.:
    N0777356
Specifications
About this item
This PC³-LT Peltier cooled inlet system for organics applications, which includes a cyclonic spray chamber, PFA-ST nebulizer, and quartz injector, is used with Optima™ 2X00 DV, 3X00 DV, 4X00 DV, 5X00 DV, 7X00 DV, and 8X00 ICP-OES series instruments.
The PC³ is a small, robust Peltier cooled cyclonic spray chamber. This offers the advantages of low memory effects, constant temperature, fast rinse-out, and high sample transport efficiency. The cooled outer walls of the PC³ spray chamber enhance long-term signal stability, reduce polyatomic interferences such as oxides, and reduce solvent loading when volatile organic solvents are analyzed. This has two preset temperatures +2 °C / -5 °C and is ideal for analysis of aqueous or aqueous/organic mixtures. The PC³-LT operates at +2 °C / -20 °C to reduce the solvent load on the plasma for analysis of 100% volatile organic solvents.
Delivery: This sample introduction kit includes: PC³-LT Peltier Cooler, Power Supply PC³-LT Baffled Quartz Cyclonic Spray Chamber with 90° Exit Connection to ESI Injector, Drain Line Adapter for Injector, PFA-ST MicroFlow Nebulizer, 0.5 mm I. Ø. ST Nebulizer Capillary, 0.25 mm I. Ø. ST Nebulizer Capillary, 1 mm I. Ø. Convertible Quartz Injector, Three Piece Mounting Shelf, 0.76 mm I. Ø. Solva Pump Tubing (4), 1.14 mm I.Ø. Solva Pump Tubing (4).