About this item
If you desire high-speed and comfort in your manual microtomy, the HistoCore BIOCUT could be the perfect solution for you.
- Quickly and efficiently select the turn direction most comfortable for you with the personalized coarse feed wheel
- Shorten cleaning time from minutes to seconds with the antistatic waste tray
- Switch between high and low-profile blades even faster without changing blade holders by using the 2-in-1 blade holder
With new options such as the personalized coarse feed wheel and 2-in-1 blade holder, it ranks among the most ergonomically friendly microtomes on the market.
Operating humidity: 20 to maximum 80% non-condensing
Operating temperature: 18...35 °C