NIR Quartz SUPRASIL® 300 Macro Cylindrical Cell with PTFE Stopper, PerkinElmer
Supplier: PerkinElmer
Certificates
About this item
The NIR quartz SUPRASIL® 300 macro cylindrical cell are used with spectrophotometers that have a larger circular light beam.
Each cell has an outside diameter of 22 mm and an inside diameter of 19 mm.
Packaging: Each package contains one cell and two windows.