Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

UV Wavecal Solution, PerkinElmer

UV wavecal solution, 500 ml
Catalog # 97012-032
Supplier:   PerkinElmer
 :  N0582152
UV Wavecal Solution, PerkinElmer
Catalog # 97012-032
Supplier:  PerkinElmer
CAS Number:  
UV Wavecal Solution, PerkinElmer
UV Wavecal Solution, PerkinElmer

UV wavecal solution, 500 ml

CAS Number:  
Price
$121.33
Log in for availability
Each (500ml)
 
Product Details & Documents

100 ppm K, P, S; 20 ppm As, La, Li, Mn, Mo, Na, Ni, Sc; 1 ppm Ca; 5% HCL.

From instrument installation to instrument calibration, we provide quality set-up, check and calibration standards to ensure instrument performance.

Specifications

Specifications

  • Description:
    UV wavecal solution
  • Size:
    500 ml

Specifications