D-Torch 3-Slot Quartz Outer Tube for Optima™ 4300/5300/7300 V, PerkinElmer
Catalog # 10136-424
Supplier: PerkinElmer
Specifications
- Description:D-Torch 3-slot quartz outer tube for Optima™
- Cat. No.:10136-424
- Supplier no.:N0777053
Specifications
About this item
Quartz outer tube for D-Torch™ used with Optima™ 4300, 5300 and 7300 V ICP-OES series instruments.
The D-Torch is a revolutionary new demountable torch design with interchangeable outer tubes. Quartz outer tubes are ideal for most aqueous samples, while Ceramic Outer Tubes (101677-618) may last longer for samples with high dissolving solids or for organic applications, such as wear metals in engine oils. This 3-slot demountable quartz outer tube allows for easy replacement and maintenance of the D-Torch. The replaceable outer tube is also more economical to replace than having to change the complex quartz assembly of the standard Optima™ torch.
Delivery: Only includes outer tube, not complete D-Torch assembly.