ContactUsLinkComponent
Rectangular Zinc Selenide Sealed Cell, PerkinElmer
Catalog # 10136-366
Supplier:  PerkinElmer
undefined
Rectangular Zinc Selenide Sealed Cell, PerkinElmer
Catalog # 10136-366
Supplier:  PerkinElmer
Supplier Number:  L1270970
Specifications
  • Description:
    Rectangular zinc selenide sealed cell
  • Cat. No.:
    10136-366
  • Pathlength:
    0.1 mm
  • Supplier no.:
    L1270970
Specifications
About this item
Sealed liquid cells are designed as a preassembled integral unit. They are ideal for highly volatile samples and quantitative analysis.
  • Made of zinc selenide
  • Pathlength of 0.1 mm
The sealed liquid cell has a corrosion-resistant construction and a precise pathlength which is calibrated using the interference pattern generated by an empty cell in the beam of a spectrometer.