Order Entry
Puerto Rico
ContactUsLinkComponent
illustra MicroSpin G-25 Columns, Cytiva
illustra MicroSpin G-25 Columns, Cytiva
  95017-621
 :  Cytiva
Product available on GSA Advantage®
 :  
undefined
illustra MicroSpin G-25 Columns, Cytiva
  95017-621
 :  Cytiva
 :  27-5325-01
Product available on GSA Advantage®
 :  

 

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    illustra MicroSpin G-25 Columns
  • Size:
    50 Preps
  • Sample Size:
    150 µL
  • Description:
    illustra MicroSpin G-25 Columns
  • Cat. No.:
    95017-621
  • Supplier no.:
    27-5325-01

 

 

For rapid buffer exchange/desalting of PCR products and other DNAs in a volume of 10 to 100 μl using spin column chromatography.

  • Prepacked with Sephadex™ G-25 DNA Grade and pre-equilibrated in distilled water containing 0,05% Kathon CG/ICP Biocide
  • Ready to use: requires less than 4 minutes from sample application to collection of purified product
  • Tested in nickase, single and double-stranded exonuclease and RNase assays
  • Can also be used for desalting/buffer exchange of DNA and removal of unincorporated radionucleotides from end-labelled oligonucleotides (at least 10 bases in length) in a volume of 10 to 100 μl