Order Entry
Puerto Rico
ContactUsLinkComponent
 
HemoCue® Hb 801 System, CLIA-Waived
Catalog # 76320-024
Supplier:  Hemocue
undefined
HemoCue® Hb 801 System, CLIA-Waived
Catalog # 76320-024
Supplier:  Hemocue
Supplier Number:  121920

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Measurement Range:
    1.0 - 25.6 g/dl (10 - 256 g/L, 0.62 - 15.9 mmol/L)
  • Sample Volume:
    10 µl of capillary or venous blood
  • Description:
    Hemocue® HB 801 analyzer, CLIA-waived
  • Measurement Time:
    <1 s
  • Operating Temperature:
    10...40 °C (50...104 °F)
  • Cat. No.:
    76320-024

Specifications

About this item

The HemoCue® Hb 801 system seamlessly integrates point-of-care hemoglobin testing into your workflow.

  • Results in under one second
  • Reagent-free cuvettes, improving open-vial stability
  • Large display with intuitive on-screen symbols
  • Internal quality control self-test
  • Customize your analyzer with HemoCue® 801 connect software featuring bluetooth

A complimentary download of the HemoCue® 801 connect software is all it takes to optimize your workflow even more with our new HemoCue® Hb 801 system.

The HemoCue® 801 connect software allows you to customize your analyzer to your workflow, with features such as: Configure analyzer(s) wirelessly via bluetooth or with data cable. Customizable cleaning and QC reminders, with documentation, based on your own standard procedures. Copy and paste patient results to an electronic medical record. Analyze hemoglobin trends in your practice with easy-to-use reporting tools. Access the most up-to-date user manual and package insert from HemoCue® in one convenient place.

Certifications: CLIA waived.
Ordering information: Optional liquid controls are available; contact your Avantor representative for more information.