Order Entry
Puerto Rico
ContactUsLinkComponent
Scissors, Ergonomic Standard Blade, Excelta Corp®
Scissors, Ergonomic Standard Blade, Excelta Corp®
Catalog # 75880-914
Supplier:  Avantor
Scissors, Ergonomic Standard Blade, Excelta Corp®
Catalog # 75880-914
Supplier:  Avantor

Specifications

  • Description:
    Scissors, kevlar, 2 star quality
  • Length:
    6" (150 mm)
  • Material:
    Stainless steel AISI 440
  • Curvature:
    Straight
  • Cat. No.:
    75880-914
  • Blade length:
    44 mm (1¾")
  • Supplier no.:
    299D-G

Specifications

About this item

These heavy duty general purpose scissors features ergonomic design of the handles.

  • Heavy duty scissor for general use
  • Cushioned grips, dual leaf springs

With added return springs and padded grips, these Scissors can be used for long periods of time without fatigue.