Order Entry
Puerto Rico
ContactUsLinkComponent
Rotopic™ Replacement Vacuum Pump, Excelta®
Rotopic™ Replacement Vacuum Pump, Excelta®
Catalog # 75880-670
Supplier:  Excelta
undefined
Rotopic™ Replacement Vacuum Pump, Excelta®
Catalog # 75880-670
Supplier:  Excelta
Supplier Number:  2000V

Specifications

  • Description:
    Rotopic™ replacement vacuum pump
  • Cat. No.:
    75880-670
  • Supplier no.:
    2000V

Specifications

About this item

The 2000V is a Rotopic™ replacement vacuum pump.

  • Pump generates up to 10" of mercury
  • Supplied complete with a 1/8" adaptor