ContactUsLinkComponent
pH and DO Probe Rack, Chemglass
  76333-468
 :  Chemglass
undefined
pH and DO Probe Rack, Chemglass
  76333-468
 :  Chemglass
 :  CLS-1455-R1
  • Description:
    Ph and do probe rack
  • Cat. No.:
    76333-468
Freestanding rack can hold 36 probes (pH or DO) up to 235 mm long. Rack is constructed of polypropylene with stainless steel uprights. Footprint of rack is 10.5L×4.5W".