ContactUsLinkComponent
Sampling Pipes, Harvest Tubes, Large Diameter, M18, for Bioreactors 2 - 5 L, Chemglass
Sampling Pipes, Harvest Tubes, Large Diameter, M18, for Bioreactors 2 - 5 L, Chemglass
Catalog # 76333-450
Supplier:  Chemglass
undefined
Sampling Pipes, Harvest Tubes, Large Diameter, M18, for Bioreactors 2 - 5 L, Chemglass
Catalog # 76333-450
Supplier:  Chemglass
Supplier Number:  CLS-1380-70
Specifications
  • Description:
    Large diameter sampling tube/harvest tube, m18, for 2-5 Liter bioreactors
  • Cat. No.:
    76333-450
Specifications
About this item
Pipe will fit a 2 to 5 liter reactor and has a lower diameter of ¹/₂" and a length of 212 mm into the reactor. The serrated hose connection is also ¹/₂" (12.7 mm) OØ. Can also be used as a quick way to harvest material due to its size.