Order Entry
Puerto Rico
ContactUsLinkComponent
CHEMRXNHUB™ Dual Support Stands, Benchtop Reactors, Chemglass
CHEMRXNHUB™ Dual Support Stands, Benchtop Reactors, Chemglass
Catalog # 76333-364
Supplier:  Chemglass
undefined
CHEMRXNHUB™ Dual Support Stands, Benchtop Reactors, Chemglass
Catalog # 76333-364
Supplier:  Chemglass
Supplier Number:  CG-1949-X-20

Specifications

  • Description:
    Chemrxnhub™ dual support stand only
  • Dimensions:
    22W×17.25D×43.25"H
  • Cat. No.:
    76333-364
  • Weight:
    68 lbs. (stand)

Specifications

About this item

Supports 2 reactors side by side from 300 ml to 5 L. This unique stationary reactor stand is designed to fit into most benchtop hoods.

  • Stainless steel uprights and heavy duty PTFE coated aluminum fittings provide maximum strength and chemical durability
  • Supports two reactors side by side from 300 ml to 5 L
  • Removable polyethylene spill tray
  • Adjustable middle support bracket is vertically adjustable: It accepts our 60,100 and 150 mm S.S. support clamps
  • Adjustable top support bracket for each overhead motor is vertically adjustable and accommodates most brands of overhead stirrers
  • Mounting clamps accept overhead stirrers with a MAX support arm of ⁵/₈" diameter
  • Unique design allows easy removal/installation of glassware
  • Compact footprint

ChemRxnHub dual stand can hold two reactors from 300 ml to 5 L. The stand provides support for the reaction vessel(s), glassware, overhead stirring motor, and temperature monitor.

Delivery: Stand includes removable spill tray.