Borosilicate Glass Sample Tubes, PerkinElmer
Catalog # 97019-148
Supplier: PerkinElmer
Specifications
- Description:Glass sample tubes
- Environmentally Preferable:
- Cat. No.:97019-148
- Supplier no.:L4071594
Specifications
About this item
Empty borosilicate glass sample tubes without caps for industrial hygiene applications using PerkinElmer thermal desorption (TD) instruments.
The tubes are a component in the workplace air monitoring industrial hygiene Application Kit (N6100448) for thermal desorption instruments.