Autosampler Sample Pans, Aluminum, PerkinElmer
Supplier: PerkinElmer
Certificates
About this item
Autosampler sample pans are used to prepare sample materials for PerkinElmer DSC Autosamplers. These pans are sealed and may also be used for non-robotic systems.
- Pans have an operating range down to –170 °C
- Pans can withstand a pressure of 1 bar
- Pan thickness is 0.1 mm
These aluminum sample pans can also be used with crimper (10137-514) and require cover (97012-264).