Demountable Quartz Injector, PerkinElmer
Catalog # 10136-462
Supplier: PerkinElmer
Specifications
- Description:Demountable quartz injector, 2.0 mm IØ
- Cat. No.:10136-462
- Supplier no.:N0777457
Specifications
About this item
This Quartz Injector Cassette Mount is designed for use with demountable torches in PerkinElmer NexION® 300/350 ICP-MS series instruments.
- 2.0 mm inner diameter
- Aassembly base is made from PFA and includes O-Rings
The demountable design allows the injector to be removed and replaced efficiently without dismounting the torch.