Solid Closed Screw Cap with PV Lining, FireflySci
Solid closed screw cap with PV lining, White
Catalog # 76016-672
Supplier: FIREFLYSCI, INC.
Supplier #: SC1W
Jump to:
Product Details & Documents
Closed screw cap comes with PTFE septa.
- Caps are made of polypropylene with poly-vinyl liner
- Good for mild acids, alkalis, solvents, alcohols, oils, and aqueous products
Caps may be used with a broad range of FireflySci cuvettes.
Specifications
- Color:White
- Description:Solid closed screw cap with PV lining