NIST 1930 VIS Photometric Accuracy Neutral Density Kit, FireflySci
Catalog # 76016-844
Supplier: FIREFLYSCI, INC.
CAS Number:
Specifications
- Description:NIST 1930 VIS photometric accuracy neutral density kit (1% T, 3% T, 50% T)
- Cat. No.:76016-844
- Tolerance:HF-0.3 (±0.0020 au), HF-1.5 (±0.0054 au), HF-2 (±0.0055 au)
Specifications
About this item
Firefly's equivalent of the NIST SRM 1930 calibration reference kit for testing photometric accuracy in the VIS range (400 to 700 nm).
- Photometric accuracy verification in the VIS range (400 to 700 nm)
- External dimensions of individual filters: 12.5×12.5×45 mm
- Scratch-resistant coating protects optical surfaces
- Can be used in any spectrophotometer
- Surpasses NIST-1930 requirements and tolerances
- NIST-compliant
This kit can be used in any spectrophotometer, and never requires replacement or recalibration if used properly.
Delivery: Kit includes the following filters: HF-2.0 (1%T), HF-1.5 (3%T), HF-0.3 (50%T), A30 blank holder, and case P4.