Micro IV Single-gas Detectors, GfG Instrumentation
Supplier: GFG INSTRUMENTATION, INC.
Certificates
About this item
The Micro IV single-gas detector is compact, robust and very easy to use. It is designed to be used in the harshest environments.
- Highly configurable smart sensor design
- Full 3-year warranty on O₂, CO and H₂S sensors
- Event/data logging standard
- Three programmable ceiling, STEL and TWA alarms
- AutoCal® automatic calibration
It includes a set of rubber boots to withstand hard knocks and drops.
It keeps gas detection simple. Most day-to-day functions are controlled with a single button, and calibration is fully automatic and delivers up to 3000 hours of continuous operation from a single off the shelf AA alkaline battery.
It carries a lifetime warranty on non-consumable components and two or more years of warranty on most sensors.
Certifications: UL certified.
Accessories information: Optional accessories that are available include sample draw pump, docking station, hand aspirated pump, and data download station.
Ordering information: For available accessories or additional information, contact your Avantor representative and see the attached datasheet and manual.
Delivery: Standard accessories include Micro IV detector, specified sensor, alkaline battery, rubber boot, and manual.
Value kit configuration includes (if value kit is specified) all standard accessories, calibration adapter, calibration gas, fixed flow regulator, and ABS carrying case.
Value kit configuration includes (if value kit is specified) all standard accessories, calibration adapter, calibration gas, fixed flow regulator, and ABS carrying case.