Order Entry
Puerto Rico
ContactUsLinkComponent
VeriSpec® ICP Multi-Element Trace Metals Calibration Standard, RICCA Chemical Company
VeriSpec® ICP Multi-Element Trace Metals Calibration Standard, RICCA Chemical Company
Catalog # RCRV010719100N
Supplier:  Ricca Chemical
CAS Number:  
undefined
VeriSpec® ICP Multi-Element Trace Metals Calibration Standard, RICCA Chemical Company
Catalog # RCRV010719100N
Supplier:  Ricca Chemical
Supplier Number:  RV010719-100N
CAS Number:  

Specifications

  • Matrix:
    10% HNO₃/tr HF
  • Pack type:
    Natural LDPE bottle
  • Size:
    100 ml
  • Cat. No.:
    RCRV010719100N
  • Supplier no.:
    RV010719-100N

Specifications

About this item

15 component standard.

500 ppm: Al
250 ppm: V
100 ppm: As, Be, Co, Cr, Cu, Fe, Mn, Ni, Pb, Zn
25 ppm: Cd, Se
5 ppm: Hg

Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility