ContactUsLinkComponent
VeriSpec® ICP Multi-Element QC Standard 7, RICCA Chemical Company
  RCRV010705100N
 :  
undefined
VeriSpec® ICP Multi-Element QC Standard 7, RICCA Chemical Company
  RCRV010705100N
 :  RV010705-100N
 :  
  • Matrix:
    5% HNO₃
  • Pack type:
    Natural LDPE bottle
  • Description:
    VeriSpec® ICP multi-element QC standard 7
  • Size:
    100 ml
  • Cat. No.:
    RCRV010705100N
  • Supplier no.:
    RV010705-100N
7 component standard.
1000 ppm: K, 500 ppm Si; 100 ppm Al, B, Ba, Na
50 ppm: Ag
 : Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility