Order Entry
Puerto Rico
ContactUsLinkComponent
 
VeriSpec® ICP Multi-Element Varian Calibration Standard, RICCA Chemical Company
Catalog # RCRV010653100N
Supplier:  Ricca Chemical
CAS Number:  
undefined
VeriSpec® ICP Multi-Element Varian Calibration Standard, RICCA Chemical Company
Catalog # RCRV010653100N
Supplier:  Ricca Chemical
Supplier Number:  RV010653-100N
CAS Number:  

Specifications

  • Matrix:
    10% HNO₃/tr HF
  • Pack type:
    Natural LDPE bottle
  • Description:
    VeriSpec® ICP multi-element varian calibration standard
  • Size:
    100 ml
  • Cat. No.:
    RCRV010653100N
  • Supplier no.:
    RV010653-100N

Specifications

About this item

25 components standard.

1000 ppm: Ca, Fe, K, Mg, Na
10 ppm: Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Se, Th, TI, U, V, Zn

Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility