To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- CAS No.:9007-34-5
- Source:
- Species:
- Size:1 Kit
- Environmentally Preferable:
- Protein/Peptide Name:Methacrylated Collagen
- Formulation:with Ruthenium
- Cat. No.:103792-912
Specifications
About this item
PhotoCol®-RUT is a methacrylated Type I collagen kit. These 3D collagen gels can be prepared at various concentrations and photocrosslinked (400-450nm) to provide various gel stiffness. Each kit contains 100mg of methacrylated collagen, ruthenium photoinitiator, acetic acid and a neutralization solution.
- Methacrylated Collagen Kit
- Tunable stiffness hydrogels
- Highly purified and characterized collagen
Advanced BioMatrix offers PhotoCol®, a purified methacrylated Type I bovine collagen kit. PhotoCol® provides native-like 3D collagen gels with the unique attributes to be prepared at various concentrations and crosslinked to provide various gel stiffness.The PhotoCol® kit consists of purified methacrylated Type I bovine collagen as the core component with other support reagents in the kit.