Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Methacrylated Collagen

Methacrylated Collagen

1 Kit
Catalog # 103792-910
Marketsource
Supplier:   Advanced Biomatrix
Supplier #:  5270-1KIT
Methacrylated Collagen
Catalog # 103792-910
Supplier:  Advanced Biomatrix
Marketsource
CAS Number:  
Methacrylated Collagen
Methacrylated Collagen

1 Kit

CAS Number:  
Price
$470.77
Log in for availability
Kit of 1
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

PhotoCol®-LAP is a methacrylated Type I collagen kit.These 3D collagen gels can be prepared at various concentrations and photocrosslinked (405nm) to provide various gel stiffness. Each kit contains 100 mg of methacrylated collagen, LAP photoinitiator, acetic acid and a neutralization solution.

  • Methacrylated Collagen Kit
  • Tunable stiffness hydrogels
  • Highly purified and characterized collagen

Advanced BioMatrix offers PhotoCol®, a purified methacrylated Type I bovine collagen kit. PhotoCol® provides native-like 3D collagen gels with the unique attributes to be prepared at various concentrations and crosslinked to provide various gel stiffness.

The methacrylated Type I collagen is produced from telo-peptide intact bovine collagen where the collagen has been modified by reacting the free amines, primarily the ε-amines groups of the lysine residues as well as the a-amines groups on the N-termini. > 20% of the total lysine residues of the collagen molecule have been methacrylated. The collagen is extracted from bovine hide and contains a high monomer content.

The collagen starting material was isolated from a closed herd and purified using controlled manufacturing processes. A bottle of 20 mM acetic acid solution is provided to solubilize the lyophilized methacrylated collagen at concentrations ranging from 3 to 8 mg/ml. The neutralization solution consists of an alkaline 10X phosphate buffered saline (PBS) solution which provides physiological salts and pH in the final mixture.

The photoinitiator consists of LAP to be formulated in 1X cell culture media or PBS which allows blue light photocrosslinking of the collagen at 405 nm.

Caution: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.

Specifications

Specifications

  • Size:
    1 Kit
  • Cat. No.:
    103792-910
  • Formulation:
    with LAP
  • Protein/Peptide Name:
    Methacrylated Collagen
  • CAS No.:
    9007-34-5

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.