ContactUsLinkComponent
Whatman™ Sealing Film and Foil for Microplates, Whatman products (Cytiva)
Whatman™ Sealing Film and Foil for Microplates, Whatman products (Cytiva)
Catalog # 28317-478
Supplier:  Cytiva
Product available on GSA Advantage®
undefined
Whatman™ Sealing Film and Foil for Microplates, Whatman products (Cytiva)
Catalog # 28317-478
Supplier:  Cytiva
Supplier Number:  7704-0001
Product available on GSA Advantage®
Specifications
  • Color:
    Clear
  • Description:
    Thin Cold Sealing Film with Adhesive Backing
  • Material:
    Polyester
  • Thickness:
    50 µm
  • Cat. No.:
    28317-478
  • Supplier no.:
    7704-0001
Specifications
About this item
Seals are used to control humidity and reduce evaporation of samples. They prevent spills and contamination.
  • Cold seals in clear polyester or PP are self-sticking with inert adhesive
  • Heat seals are suitable for PP microplates only and are applied using heat and pressure