Hyaluronic acid sodium salt methacrylated kit with LAP, PhotoHA®-LAP
103792-918
:
- Size:1 kit
- Cat. No.:103792-918
- Sterilization Method:Filtration
- Sterility - USP modified:No growth
- Form:Lyophilized Powder
- Package Size:100 mg
- Storage Temperature:-20°C
- Shelf Life:Minimum of 6 months from date of receipt
- Shelf Life After Reconstitution:1 Month
- Degree of Methacrylation:45-65%
- Molecular Weight:100-150 kDa
- NMR Analysis:Characteristic
- Hydrogel Young's Modulus E (Pa):Characteristic
:
:Freezer
:9067-32-7
:(C₁₄H₂₁NaNO₁₁)n