WHEATON® Open Top Sample Vials In Lab File, DWK Life Sciences
Catalog # 14221-606
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Cap included:Yes
- Cap Size:24-400
- Cap type:Lined caps
- Capacity:20.00 ml
- Color:Amber
- Cat. No.:14221-606
- Supplier no.:W227356
Specifications
About this item
These vials are manufactured from high-quality glass that meets ASTM standards.
- Amber finish
- Septa are bonded to the cap providing a permanent connection
Glass meets USP Type 1 standards to protect light sensitive samples and maintain purity of contents. The white polypropylene open top screw cap has a specially designed septum of 5 mils of Teflon facing 0.120 mils of silicone. The polypropylene is glass-filled to provide extra chemical and heat resistance compared to standard polypropylene.