WHEATON® Septa for Open Top Caps, Autoclavable, DWK Life Sciences
Catalog # 89050-893
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Ø:24 mm
- Septum:PTFE faced semi-translucent silicone 10/90
- Cat. No.:89050-893
- Supplier no.:W240598A
Specifications
About this item
These caps are autoclavable.
- For use with open top caps
Caps for locking flange septa and plain septa.