ContactUsLinkComponent
Autosampler Sample Pans, 30 µl, PerkinElmer
Autosampler Sample Pans, 30 µl, PerkinElmer
Catalog # 10136-334
Supplier:  PerkinElmer
Autosampler Sample Pans, 30 µl, PerkinElmer
Catalog # 10136-334
Supplier:  PerkinElmer
Supplier Number:  B0169320
Specifications
  • Description:
    Autosampler sample pans, 30 µl
  • Cat. No.:
    10136-334
  • Supplier no.:
    B0169320
Specifications
About this item
The Autosampler sample pans are used to prepare sample materials for PerkinElmer DSC Autosampler. These pans have an operating range of –170 to 600 ° C, a volume of 30 µl, and 3 bar maximum pressure.
  • Used by the universal crimper press (10137-514)
  • Can be used with all autosamplers
These sample pans are for use with Diamond DSC, Pyris 1 DSC, DSC 7, DSC 6, Pyris 6 DSC, and Jade DSC. These pans are hermetically sealed and may also be used for non-robotic systems.
Delivery: Includes 400 sample pans.