ContactUsLinkComponent
Sapphire Injector for Demountable ZipTorch for PerkinElmer Optima ICP-OES Instruments, PerkinElmer
Sapphire Injector for Demountable ZipTorch for PerkinElmer Optima ICP-OES Instruments, PerkinElmer
Catalog # 10136-390
Supplier:  PerkinElmer
undefined
Sapphire Injector for Demountable ZipTorch for PerkinElmer Optima ICP-OES Instruments, PerkinElmer
Catalog # 10136-390
Supplier:  PerkinElmer
Supplier Number:  N0777044
Specifications
  • Description:
    Sapphire injector for demountable ZipTorch for Optima, 1.8 mm I.D.
  • Cat. No.:
    10136-390
  • Supplier no.:
    N0777044
Specifications
About this item
This demountable injector is for use with Optima® 4x00/5x00/7x00 ICP-OES series instruments.
It is designed for use with the quartz, organics and silicon nitride demountable ZipTorches. The demountable design allows the injector to be removed and replaced efficiently for cleaning without dismounting the torch.